Learn More
ATCC Quantitative Synthetic; Pneumocystis jirovecii DNA
Supplier:  ATCC MYA5006SD
Encompass
Quantitative Synthetic Pneumocystis jirovecii DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load. This preparation includes fragments from the mtLSU rRNA, mtSSU rRNA, DHPS, MSG, KEX-1, and Beta-tubulin regions.
Catalog No. 50-238-4746