Learn More
ATCC Quantitative Synthetic; Powassan virus; lineage II RNA
Catalog No. 502383324
Encompass
Quantitative synthetic Powassan virus lineage II RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the E, NS1, NS5, and 3’ UTR regions.
Catalog No. 50-238-3324 Supplier ATCC Supplier No. VR3275SD
Quantitative synthetic Powassan virus lineage II RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the E, NS1, NS5, and 3’ UTR regions.