Learn More
ATCC Quantitative Synthetic; Severe acute respiratory; syndrome-related coronavirus 2; (SARS-CoV-2) RNA: nsp9,; nsp12 (RdRp)
Catalog No. 502381788
Encompass
Quantitative synthetic SARS-CoV-2 RNA: nsp9, nsp12 (RdRp) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the nsp9 and nsp12 (RdRp) regions.
Catalog No. 50-238-1788 Supplier ATCC Supplier No. VR3279SD
Quantitative synthetic SARS-CoV-2 RNA: nsp9, nsp12 (RdRp) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the nsp9 and nsp12 (RdRp) regions.