Learn More
ATCC Quantitative Synthetic; Severe acute respiratory; syndrome-related coronavirus; (SARS-CoV-2) RNA: Spike 3'
Catalog No. 502382356
Encompass
Quantitative synthetic SARS-CoV-2 RNA: Spike 3' can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes a fragment from the 3’ glycoprotein (Spike) region.
Catalog No. 50-238-2356 Supplier ATCC Supplier No. VR3278SD
Quantitative synthetic SARS-CoV-2 RNA: Spike 3' can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes a fragment from the 3’ glycoprotein (Spike) region.