Learn More
ATCC Quantitative Synthetic; Severe acute respiratory; syndrome-related coronavirus; (SARS-CoV-2) RNA: Spike 5'
Catalog No. 502382487
Encompass
Quantitative synthetic SARS-CoV-2 RNA: Spike 5' can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes a fragment from the 5’ glycoprotein (Spike) region.
Catalog No. 50-238-2487 Supplier ATCC Supplier No. VR3277SD
Quantitative synthetic SARS-CoV-2 RNA: Spike 5' can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes a fragment from the 5’ glycoprotein (Spike) region.