Learn More
ATCC Quantitative Synthetic SARS-CoV-2 RNA: ORF, E, N (ATCC VR-3276SD™)
Catalog No. 502382339
Encompass
Quantitative Synthetic SARS-CoV-2 RNA (ATCC VR-3276SD™) includes fragments from the ORF1ab, envelope (E), and nucleocapsid (N) regions. Manufactured under ISO 13485 guidance, it is used for assay development, qPCR calibration, and molecular diagnostics development.
Catalog No. 50-238-2339 Supplier ATCC Supplier No. VR3276SD
Quantitative Synthetic SARS-CoV-2 RNA (ATCC VR-3276SD™) includes fragments from the ORF1ab, envelope (E), and nucleocapsid (N) regions. Manufactured under ISO 13485 guidance, it is used for assay development, qPCR calibration, and molecular diagnostics development.