Learn More
ATCC Quantitative Synthetic; Severe acute respiratory; syndrome-related coronavirus; (SARS-CoV) [2003] RNA: nsp9;(RdRp), nsp 11, N
Catalog No. 502381778
Encompass
Quantitative synthetic SARS-CoV [2003] RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the nsp9 (RdRp), nsp11, and N regions.
Catalog No. 50-238-1778 Supplier ATCC Supplier No. VR3280SD
Quantitative synthetic SARS-CoV [2003] RNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the nsp9 (RdRp), nsp11, and N regions.