Learn More
CTL Scientific Supply Corp QUANTOFIX ALUMMINUM WITH REAGE
SDP
Catalog No. 50300356 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX ALUMMINUM WITH REAGENTS 100 strips/pack plus reagents Test strips for the semi-quantitative determination of alumminum in solutions. Gradation of scale: 0-5-20-50-200-500 ppm Al3+
Catalog No. 50-300-356 Supplier CTL Scientific Supply Corp Supplier No. 91307
QUANTOFIX ALUMMINUM WITH REAGENTS 100 strips/pack plus reagents Test strips for the semi-quantitative determination of alumminum in solutions. Gradation of scale: 0-5-20-50-200-500 ppm Al3+