Learn More
CTL Scientific Supply Corp QUANTOFIX AMMONIUM
SDP
Catalog No. 50300303 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX AMMONIUM, Test strips and reagents for the semi-quantitative determination of ammonium in solutions. 100 strips/pack Gradation of scale: 0-10-25-50-100-200-400 ppm
Catalog No. 50-300-303 Supplier CTL Scientific Supply Corp Supplier No. 91315
QUANTOFIX AMMONIUM, Test strips and reagents for the semi-quantitative determination of ammonium in solutions. 100 strips/pack Gradation of scale: 0-10-25-50-100-200-400 ppm