Learn More
CTL Scientific Supply Corp QUANTOFIX ASCORBIC ACID
SDP
Catalog No. 50300346 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test sticks for the semi-quantitative determination of vitamin C in solutions.
Gradation of scale: 0-50-100-200-300-500-1000-2000 mg/l 100 strips/box Protect from sunlight and moisture.
Catalog No. 50-300-346 Supplier CTL Scientific Supply Corp Supplier No. 91314
Test sticks for the semi-quantitative determination of vitamin C in solutions.
Gradation of scale: 0-50-100-200-300-500-1000-2000 mg/l 100 strips/box Protect from sunlight and moisture.