Learn More
CTL Scientific Supply Corp QUANTOFIX CARBONATE HARDNESS
SDP
Supplier:  CTL Scientific Supply Corp 91323
Encompass_Preferred
Test sticks for the semi-quantitative determination of the carbonate hardness. Carbonate hardness is that part of the total hardness resulting from the content of alkaline earth ions, Gradation of scale: 0-3-6-10-15-20 100 strips
Catalog No. 50-300-348