Learn More
CTL Scientific Supply Corp QUANTOFIX CHLORIDE
SDP
Catalog No. 50300347 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX CHLORIDE 100 strips/pack Test strips for the semi-quantitative determination of chloride in solutions. Gradation of scale: 0-500-1000-1500-2000-3000 ppm Cl-
Catalog No. 50-300-347 Supplier CTL Scientific Supply Corp Supplier No. 91321
QUANTOFIX CHLORIDE 100 strips/pack Test strips for the semi-quantitative determination of chloride in solutions. Gradation of scale: 0-500-1000-1500-2000-3000 ppm Cl-