Learn More
CTL Scientific Supply Corp QUANTOFIX COBALT
SDP
Catalog No. 50300344 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test sticks for the semi-quantitative determination of cobalt in solutions. Gradation of scale: 0-10-25-50-100-250-500-1000 mg/l 100 strips/box
Catalog No. 50-300-344 Supplier CTL Scientific Supply Corp Supplier No. 91303
Test sticks for the semi-quantitative determination of cobalt in solutions. Gradation of scale: 0-10-25-50-100-250-500-1000 mg/l 100 strips/box