Learn More
CTL Scientific Supply Corp QUANTOFIX CYANIDE WITH REAGENT
SDP
Catalog No. 50300359 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX CYANIDE WITH REAGENTS 100 strips/pack Test strips for the semi-quantitative determination of cyanide in solutions. Gradation of scale: 0-1-3-10-30 ppm CN-
Catalog No. 50-300-359 Supplier CTL Scientific Supply Corp Supplier No. 91318
QUANTOFIX CYANIDE WITH REAGENTS 100 strips/pack Test strips for the semi-quantitative determination of cyanide in solutions. Gradation of scale: 0-1-3-10-30 ppm CN-