Learn More
CTL Scientific Supply Corp QUANTOFIX EDTA
Supplier: CTL Scientific Supply Corp 91335
QUANTOFIX EDTA 100 strips/pack Test strips for the semi-quantitative determination of EDTA, NTA and other complexing agents in solutions. Gradation of scale: 0-100-200-300-400 ppm EDTA