Learn More
CTL Scientific Supply Corp QUANTOFIX FORMALDEHYDE
SDP
Supplier:  CTL Scientific Supply Corp 91328
Encompass_Preferred
Test sticks and reagents for the semi-quantitative determination of formaldehyde in solutions. Gradation of scale: 0-10-20-40-60-100-200 mg/l HCHO beige to blue-violet. 100 strips/box + reagents
Catalog No. 50-300-333