Learn More
CTL Scientific Supply Corp QUANTOFIX NICKEL
SDP
Catalog No. 50300302 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test strips for the semi-quantitative determination of nickel (Ni 2+ ) in solutions. 100 strips/pack Gradation of scale: 0-10-25-50-100-250-500-1000 ppm.
Catalog No. 50-300-302 Supplier CTL Scientific Supply Corp Supplier No. 91305
Test strips for the semi-quantitative determination of nickel (Ni 2+ ) in solutions. 100 strips/pack Gradation of scale: 0-10-25-50-100-250-500-1000 ppm.