Learn More
CTL Scientific Supply Corp QUANTOFIX NITRITE 3000
SDP
Catalog No. 50300310 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test sticks for the semi-quantitative determination of high concentrations of nitrite (NO 2 ) in solutions
Gradation of scale: 0-0.1-0.3-0.6-1-2-3 g/l 200 strips/box
Catalog No. 50-300-310 Supplier CTL Scientific Supply Corp Supplier No. 91322
Test sticks for the semi-quantitative determination of high concentrations of nitrite (NO 2 ) in solutions
Gradation of scale: 0-0.1-0.3-0.6-1-2-3 g/l 200 strips/box