Learn More
CTL Scientific Supply Corp QUANTOFIX NITRITE/PH FOR COOLI
SDP
Catalog No. 50300355 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test strips allowing quick and easy determination of Nitrite and pH in cooling lubricants. Easy dip-and-read-procedure provides a reliable result within 1 minute. Scale 0.1-5-10-20-40-80- mgL pH 6.0-6.4-6.7-7.0-7.6-7.9-8.2-8.4-8.6-8.8-9.0-9.3-9.6
Catalog No. 50-300-355 Supplier CTL Scientific Supply Corp Supplier No. 91338
Test strips allowing quick and easy determination of Nitrite and pH in cooling lubricants. Easy dip-and-read-procedure provides a reliable result within 1 minute. Scale 0.1-5-10-20-40-80- mgL pH 6.0-6.4-6.7-7.0-7.6-7.9-8.2-8.4-8.6-8.8-9.0-9.3-9.6