Learn More
CTL Scientific Supply Corp QUANTOFIX NITRITE
SDP
Catalog No. 50300331 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX NITRITE 100 strips/pack Test strips for the semi-quantitative determination of nitrite in solutions. Gradation of scale: 0-10-25-50-100-250-500-1000 ppm SO32-
Catalog No. 50-300-331 Supplier CTL Scientific Supply Corp Supplier No. 91311
Add to Cart
Add to Cart
QUANTOFIX NITRITE 100 strips/pack Test strips for the semi-quantitative determination of nitrite in solutions. Gradation of scale: 0-10-25-50-100-250-500-1000 ppm SO32-