Learn More
Macherey-Nagel Semi-quantitative test strips QUANTOFIX Peroxide 1000, 100 tests
Catalog No. NC1247324
Encompass_Preferred
Semi-quantitative test strips for the determination of Peroxide. Ideal for rapid and easy analysis of Peroxide directly on-site. Can be used with test strip reader QUANTOFIX Relax.
Catalog No. NC1247324 Supplier Macherey-Nagel Supplier No. 91333
Semi-quantitative test strips for the determination of Peroxide. Ideal for rapid and easy analysis of Peroxide directly on-site. Can be used with test strip reader QUANTOFIX Relax.