Learn More
CTL Scientific Supply Corp QUANTOFIX PEROXIDE 1000
SDP
Supplier:  CTL Scientific Supply Corp 91333
Encompass_Preferred
QUANTOFIX FOR PEROXIDE 100 strips/pack Test strips for the semi-quantitative determination of peroxide in solutions. Gradation of scale: 0-50-150-300-500-800-1000 ppm H2O2
Catalog No. 50-300-330