Learn More
CTL Scientific Supply Corp QUANTOFIX Peroxide Test Strips, 100/bx
SDP
Catalog No. 50300317 Shop All CTL Scientific Supply Products
Encompass_Preferred
QUANTOFIX FOR PEROXIDE Test 100 strips/pack Test strips for the semi-quantitative determination of peroxide in solutions. Gradation of scale 0-1-3-10-30-100 ppm H2O2
Catalog No. 50-300-317 Supplier CTL Scientific Supply Corp Supplier No. 91312
QUANTOFIX FOR PEROXIDE Test 100 strips/pack Test strips for the semi-quantitative determination of peroxide in solutions. Gradation of scale 0-1-3-10-30-100 ppm H2O2