Learn More
CTL Scientific Supply Corp QUANTOFIX POTASSIUM WITH REAGE
SDP
Catalog No. 50300358 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test sticks and reagents for the semi-quantitative determination of potassium (K + ) in solutions
Gradation of scale: 0-200-400-700-1000-1500 mg/l 100 strips/box + reagents
Catalog No. 50-300-358 Supplier CTL Scientific Supply Corp Supplier No. 91316
Add to Cart
Add to Cart
Test sticks and reagents for the semi-quantitative determination of potassium (K + ) in solutions
Gradation of scale: 0-200-400-700-1000-1500 mg/l 100 strips/box + reagents