Learn More
CTL Scientific Supply Corp QUANTOFIX TIN
SDP
Catalog No. 50300345 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test sticks for the semi-quantitative determination of tin(II)-ions in solutions. Gradation of scale: 0-10-25-50-100-250-500 mg/l 100 strips/box PRODUCT NOT RECOMMENDED FOR USE BY NON CHEMIST
Catalog No. 50-300-345 Supplier CTL Scientific Supply Corp Supplier No. 91309
Test sticks for the semi-quantitative determination of tin(II)-ions in solutions. Gradation of scale: 0-10-25-50-100-250-500 mg/l 100 strips/box PRODUCT NOT RECOMMENDED FOR USE BY NON CHEMIST