Learn More
CTL Scientific Supply Corp QUANTOFIX ZINC
SDP
Catalog No. 50300315 Shop All CTL Scientific Supply Products
Encompass_Preferred
Test strips used for the semi-quantitative determination of zinc in solutions. Gradation of scale: 0-2-5-10-25-30-100 ppm Zn2+ - 100 strips/pack plus reagents.
Catalog No. 50-300-315 Supplier CTL Scientific Supply Corp Supplier No. 91310
Test strips used for the semi-quantitative determination of zinc in solutions. Gradation of scale: 0-2-5-10-25-30-100 ppm Zn2+ - 100 strips/pack plus reagents.