Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Quantos Dosing Heads

Catalog No. 01910260 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Application):
Compacting powders
Difficult to dose powder
Mining substances and beads
Recommended 250 dosings up to 500 mg
Several grams target weight
Small target container
Size (Metric):
2.5 mm
4 mm
Volume (Metric):
8 mL
12 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Size (Metric) Volume (Metric)
01-910-260 Difficult to dose powder 4 mm 12 mL
01-910-259 Compacting powders 4 mm 12 mL
01-910-258 Several grams target weight 4 mm 12 mL
01-910-256 Recommended 250 dosings up to 500 mg 2.5 mm 8 mL
01-910-254 Recommended 250 dosings up to 500 mg 2.5 mm 12 mL
01-910-262 Mining substances and beads 4 mm 12 mL
01-910-261 Small target container 2.5 mm 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-910-260 Supplier Mettler Toledo™ Supplier No. 11150171
Add to Cart
Edge
Add to Cart

Intelligent dosing heads for precise dosing of free flowing powdery substances such as APIs or reference standards where purity and quality of substance is essential. Mettler Toledo Quantos Dosing Head includes an integrated stirrer which guarantees a homogeneous flow of the powder.

Four motors drive the powder flow with real time feedback from the balance, no settings to play with. The use of a closed system avoids cross-contamination and protects the user, the substance and the environment.

  • Composed of FDA compliant materials
  • Made of TPX polymer and 316L/1.4404 stainless steel which feature electrical, chemical and physical properties
  • Each head assigned to single substance eliminates cross-contamination
  • All substance information (substance ID,quantity, date etc.) traceable via RFID chip in each dosing head
  • Dosing head attached to container for lifetime of substance
  • No training required for using dosing head, place head on Quantos device and start working
  • Supplied in impact resistant, water-tight transport container
  • Sterile versions available

†Sterile

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Laboratory Balances
Material TPX Polymer and 316L/1.4404 Stainless Steel
Volume (Metric) 12 mL
For Use With (Application) Difficult to dose powder
Includes w/o Container
Size (Metric) 4 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.