Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Quantos Sample Heads

Catalog No. 01910265 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
Set of 30
Set of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-910-265 Set of 30
01-910-266 Set of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-910-265 Supplier Mettler Toledo™ Supplier No. 30083440
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Precise dosing; For few dispenses and low starting quantities.

  • TPX polymer
  • Integrated RFID chip
  • Small
  • 2.5mm pin
  • Max. 2mL substance volume
  • With stirrer
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Laboratory Balances
Material TPX Polymer and 316L/1.4404 Stainless Steel
Volume (Metric) Max. 2 mL
Size (Metric) 2.5 mm
Quantity Set of 30
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.