Learn More
World Precision Instrument Quantrex Ultrasonic Cleaning System
SDP
Catalog No. 50380073
Encompass
Quantrex bath with a timer
  • 60 Minute timer
  • Vinyl-clad steel and stainless steel
  • 14 quality inspection steps for strength and durability
  • Includes timer, drain and cover
  • Heater included
  • Sold in the USA and Canada ONLY
Catalog No. 50-380-073 Supplier World Precision Instrument Supplier No. 504216
Quantrex bath with a timer
  • 60 Minute timer
  • Vinyl-clad steel and stainless steel
  • 14 quality inspection steps for strength and durability
  • Includes timer, drain and cover
  • Heater included
  • Sold in the USA and Canada ONLY