Learn More
Lexogen QuantSeq-Pool Sample-Barcoded 3′ mRNA-Seq Library Prep Kit for Illumina
SDP
Catalog No. NC1857055
Encompass
QuantSeq offers an all-in-one library preparation protocol designed to generate Illumina compatible libraries of sequences close to the 3’ end of polyadenylated RNA. QuantSeq-Pool is a streamlined solution for gene expression profiling for large screening projects using early pooling and batch processing and is easily scalable from a few to 36,864 samples.
Catalog No. NC1857055 Supplier Lexogen Supplier No. 139.96
QuantSeq offers an all-in-one library preparation protocol designed to generate Illumina compatible libraries of sequences close to the 3’ end of polyadenylated RNA. QuantSeq-Pool is a streamlined solution for gene expression profiling for large screening projects using early pooling and batch processing and is easily scalable from a few to 36,864 samples.