Learn More
BANGS LABORATORIES INC Bangs Quantum™ Alexa Fluor 647 MESF Kit, Fluorescence Standardization, 100 Tests
Catalog No. NC3628523
Encompass
The Bangs Laboratories Quantum™ Alexa Fluor 647 MESF 5mL kit is essential for quantitative fluorescence cytometry. It standardizes fluorescence intensity units through a calibration curve correlating instrument values with MESF units. Its key benefits include accurate cellular expression analysis and evaluation of instrument performance, with microspheres labeled for environmental responsiveness.
Catalog No. NC3628523 Supplier BANGS LABORATORIES INC Supplier No. 647B
The Bangs Laboratories Quantum™ Alexa Fluor 647 MESF 5mL kit is essential for quantitative fluorescence cytometry. It standardizes fluorescence intensity units through a calibration curve correlating instrument values with MESF units. Its key benefits include accurate cellular expression analysis and evaluation of instrument performance, with microspheres labeled for environmental responsiveness.