Learn More
BANGS LABORATORIES INC Quantum Alexa Fluor 488 MESF Kit, Quantitative Flow Cytometry Standardization, QuickCal Analysis Template, 5 mL
Catalog No. NC0397712
Encompass
The Bangs Laboratories Quantum Alexa Fluor 488 MESF kit offers precise calibration for flow cytometry, enhancing accuracy in measuring cell expression levels. This quantitative MESF kit contains a blank and four distinct fluorescent microsphere populations, ideal for standardizing instrument performance. With relevant size approximation to human lymphocytes, it ensures superior applicability in research and diagnostics. Includes a QuickCal analysis template for simplified data interpretation.
Catalog No. NC0397712 Supplier BANGS LABORATORIES INC Supplier No. 488 5ML B
Add to Cart
Add to Cart
The Bangs Laboratories Quantum Alexa Fluor 488 MESF kit offers precise calibration for flow cytometry, enhancing accuracy in measuring cell expression levels. This quantitative MESF kit contains a blank and four distinct fluorescent microsphere populations, ideal for standardizing instrument performance. With relevant size approximation to human lymphocytes, it ensures superior applicability in research and diagnostics. Includes a QuickCal analysis template for simplified data interpretation.