Learn More
BANGS LABORATORIES INC Quantum Alexa Fluor 647 MESF Kit, Standardized Fluorescence Intensity, 100 Tests, 5 mL
Catalog No. NC0403002
Encompass
The Quantum™ Alexa Fluor 647 MESF 5mL kit by Bangs Laboratories empowers precision in flow cytometry, providing standardized fluorescent microspheres crucial for accurate calibration and data consistency. With five populations for diverse experimental assessments and a focus on enabling reproducible fluorescence quantitation, researchers can effectively gauge instrument performance and fluorochrome expression levels.
Catalog No. NC0403002 Supplier BANGS LABORATORIES INC Supplier No. 647B5ML
The Quantum™ Alexa Fluor 647 MESF 5mL kit by Bangs Laboratories empowers precision in flow cytometry, providing standardized fluorescent microspheres crucial for accurate calibration and data consistency. With five populations for diverse experimental assessments and a focus on enabling reproducible fluorescence quantitation, researchers can effectively gauge instrument performance and fluorochrome expression levels.