Learn More
LOGOS BIOSYSTEMS INC QUANTOM™ M50 Cell Counting Slides, 50 Slides
SDP
Catalog No. NC1774374
Encompass
QUANTOM™ M50 Cell Counting Slides are disposable slides designed for use with the QUANTOM Tx™. The disposable QUANTOM™ M50 Cell Counting Slides are made with optical quality materials with ultra-low autofluorescence to allow for optimal fluorescence detection. QUANTOM™ M50 Cell Counting Slides maintain the highest standard of cell counting accuracy and offer the ultimate counting experience with no mess or cleanup.
Catalog No. NC1774374 Supplier LOGOS BIOSYSTEMS INC Supplier No. Q12001
QUANTOM™ M50 Cell Counting Slides are disposable slides designed for use with the QUANTOM Tx™. The disposable QUANTOM™ M50 Cell Counting Slides are made with optical quality materials with ultra-low autofluorescence to allow for optimal fluorescence detection. QUANTOM™ M50 Cell Counting Slides maintain the highest standard of cell counting accuracy and offer the ultimate counting experience with no mess or cleanup.