Learn More
BANGS LABORATORIES INC Quantum™ FITC-5 MESF Kit, Accurate Measurement of Fluorescence Intensity, Includes QuickCal Analysis Template, 1 mL
Catalog No. NC2077226
Encompass
The Quantum™ FITC-5 MESF kit provides a reliable solution for quantifying fluorescence intensity in flow cytometry. Designed for researchers needing precise FITC measurements, it aids in instrument calibration and data consistency across experiments. With microspheres mimicking human lymphocytes' size and a comprehensive analysis template, it ensures accurate cell expression evaluation and instrument performance. The kit includes 1 mL of solution, 20 tests, and features environmentally responsive microspheres.
Catalog No. NC2077226 Supplier BANGS LABORATORIES INC Supplier No. 5551ML
The Quantum™ FITC-5 MESF kit provides a reliable solution for quantifying fluorescence intensity in flow cytometry. Designed for researchers needing precise FITC measurements, it aids in instrument calibration and data consistency across experiments. With microspheres mimicking human lymphocytes' size and a comprehensive analysis template, it ensures accurate cell expression evaluation and instrument performance. The kit includes 1 mL of solution, 20 tests, and features environmentally responsive microspheres.