Learn More
BANGS LABORATORIES INC Quantum™ FITC-5 MESF, Calibrated Reference Material, Enables Quantitative Fluorescence Cytometry, 1 mL
Catalog No. NC1954535
Encompass
The Quantum™ FITC-5 MESF is a calibrated fluorescence cytometry reference kit designed to standardize and quantify FITC fluorescence intensity in MESF units. It helps assess flow cytometer performance by ensuring accurate comparisons between instruments over time. The kit uniquely includes a QuickCal analysis template for evaluating cell expression levels, differentiating it from other products by providing essential tools for precise fluorescence quantification.
Catalog No. NC1954535 Supplier BANGS LABORATORIES INC Supplier No. 555A
The Quantum™ FITC-5 MESF is a calibrated fluorescence cytometry reference kit designed to standardize and quantify FITC fluorescence intensity in MESF units. It helps assess flow cytometer performance by ensuring accurate comparisons between instruments over time. The kit uniquely includes a QuickCal analysis template for evaluating cell expression levels, differentiating it from other products by providing essential tools for precise fluorescence quantification.