Learn More
Polysciences, Inc. Quantum R-PE MESF, 1 mL
SDP
Catalog No. NC1618312
Encompass
Quantitative MESF Kits consist of a series of reference microbead standards whose intensities have been calibrated in Molecules of Equivalent Soluble Fluorochrome (MESF) units.
Catalog No. NC1618312 Supplier Polysciences, Inc. Supplier No. BLI827A1
Quantitative MESF Kits consist of a series of reference microbead standards whose intensities have been calibrated in Molecules of Equivalent Soluble Fluorochrome (MESF) units.