Learn More
BANGS LABORATORIES INC QUANTUM SIMPLY CELLULAR 5ML
Supplier:  BANGS LABORATORIES INC 815B5ML
Encompass
Quantum Simply Cellular Anti-mouse IgG Medium Level (~10,000 - 200,000 ABC), 1 bottle, size:100T, Volume/ml:5
Catalog No. NC9953428