Learn More
BANGS LABORATORIES INC Quantum™ Simply Cellular Anti-Human IgG, Quantitative Flow Cytometry Standard, 5 mL
Catalog No. NC0950894
Encompass
Quantum™ Simply Cellular anti-Human IgG is a quantitative flow cytometry standard designed to enhance accuracy in determining Antibody Binding Capacity (ABC) of cells. This product is vital for researchers aiming for consistency across various instruments and software, allowing for precise quantification of cellular antigen expression. Its unique microsphere populations facilitate calibration and direct comparisons of cell populations, distinguishing it as a reliable resource for flow cytometry applications.
Catalog No. NC0950894 Supplier BANGS LABORATORIES INC Supplier No. 816B5ML
Quantum™ Simply Cellular anti-Human IgG is a quantitative flow cytometry standard designed to enhance accuracy in determining Antibody Binding Capacity (ABC) of cells. This product is vital for researchers aiming for consistency across various instruments and software, allowing for precise quantification of cellular antigen expression. Its unique microsphere populations facilitate calibration and direct comparisons of cell populations, distinguishing it as a reliable resource for flow cytometry applications.