Learn More
MSC Quantum Storage Dividable Storage Tote, Polypropylene, 50 lb, Stacking
Catalog No. 5000534515
Encompass
Quantum Storage polypropylene dividable storage totes are designed for efficient organization and storage. They feature a durable polypropylene construction and a 50 lb load capacity, making them suitable for various industrial and commercial applications. These totes are also stackable, offering space-saving solutions.
  • Dividable container type
  • Constructed from polypropylene
  • 50 lb load capacity
  • Stackable design
  • Non-nesting
  • Gray color
  • Compatible with short and long dividers, and lids
Catalog No. 50-005-34515 Supplier MSC Supplier No. 86549722
Quantum Storage polypropylene dividable storage totes are designed for efficient organization and storage. They feature a durable polypropylene construction and a 50 lb load capacity, making them suitable for various industrial and commercial applications. These totes are also stackable, offering space-saving solutions.
  • Dividable container type
  • Constructed from polypropylene
  • 50 lb load capacity
  • Stackable design
  • Non-nesting
  • Gray color
  • Compatible with short and long dividers, and lids