Learn More
MSC Quantum Storage Drag Chain, Zinc, 17 in, For Wire Shelving Units
Catalog No. 5000527150
Encompass
This drag chain is designed for use with wire shelving units. Made from zinc with a zinc-plated finish, it provides a conductive connection for grounding applications.
  • Conductive cable connects ends
  • Used in conjunction with aluminum split sleeves
  • Shelf end of chain clips onto post
  • 17 in overall length
Catalog No. 50-005-27150 Supplier MSC Supplier No. 40660797
This drag chain is designed for use with wire shelving units. Made from zinc with a zinc-plated finish, it provides a conductive connection for grounding applications.
  • Conductive cable connects ends
  • Used in conjunction with aluminum split sleeves
  • Shelf end of chain clips onto post
  • 17 in overall length