Learn More
MSC Quantum Storage Floor Pick Rack, Double-Sided Mobile, 4200 lb Capacity, 96 Green Bins
Catalog No. 5000532426
Encompass
This mobile floor pick rack is a double-sided unit designed for efficient storage and organization, featuring a robust 4,200 lb capacity and integrated shelf bins. It provides a versatile solution for various industrial and commercial settings.
  • Double-sided mobile design
  • 4,200 lb load capacity
  • Includes 96 green polyethylene and polypropylene bins
  • Durable steel rack with powder coated finish
  • Open shelving style
  • Dimensions: 12 inch depth, 75 inch height, 36 inch width
  • 13 shelves for optimal storage
Catalog No. 50-005-32426 Supplier MSC Supplier No. 48520787
This mobile floor pick rack is a double-sided unit designed for efficient storage and organization, featuring a robust 4,200 lb capacity and integrated shelf bins. It provides a versatile solution for various industrial and commercial settings.
  • Double-sided mobile design
  • 4,200 lb load capacity
  • Includes 96 green polyethylene and polypropylene bins
  • Durable steel rack with powder coated finish
  • Open shelving style
  • Dimensions: 12 inch depth, 75 inch height, 36 inch width
  • 13 shelves for optimal storage