Learn More
MSC Quantum Storage Hopper Shelf Bin, Polypropylene, Conductive, 23-5/8 x 6-5/8 x 4 in
Catalog No. 5000530040
Encompass
This conductive polypropylene hopper shelf bin is designed for organized storage within wire shelving systems. Featuring a hopper front and label holder for easy access and identification, it is ideal for environments requiring electrostatic discharge protection. Its specialized material ensures a surface resistivity of less than 105 Ohms per square, preventing the build-up of electrostatic charges.
  • Hopper front bin and label holder
  • Surface resistivity is less than 105 Ohms-per square
  • Prevents build-up of electrostatic charges
  • Made of polypropylene material
  • Designed for wire shelving systems
  • Nesting capable
Catalog No. 50-005-30040 Supplier MSC Supplier No. 00647131
This conductive polypropylene hopper shelf bin is designed for organized storage within wire shelving systems. Featuring a hopper front and label holder for easy access and identification, it is ideal for environments requiring electrostatic discharge protection. Its specialized material ensures a surface resistivity of less than 105 Ohms per square, preventing the build-up of electrostatic charges.
  • Hopper front bin and label holder
  • Surface resistivity is less than 105 Ohms-per square
  • Prevents build-up of electrostatic charges
  • Made of polypropylene material
  • Designed for wire shelving systems
  • Nesting capable