Learn More
MSC Quantum Storage Hopper Shelf Bin, Polypropylene, Red, 50 Lb Capacity
Catalog No. 5000537053
Encompass
This plastic hopper shelf bin is constructed from durable polypropylene and designed for efficient storage. It features a 50 Lb capacity and measures 17-7/8 in L x 4-3/8 in W x 6 in H, making it suitable for various organizational needs in industrial and laboratory settings. This bin is supplied in a convenient 20-pack and is compatible with a range of shelving units.
  • Molded-in label holder for easy identification.
  • Reinforced edges for enhanced durability.
  • Waterproof and corrosion-free construction ensures longevity.
  • Durable material provides reliable storage.
  • Designed for use with 12 in, 18 in, and 24 in deep shelf units, racks, or standard shelving.
  • Nestable design allows for efficient storage when empty.
Catalog No. 50-005-37053 Supplier MSC Supplier No. 91825596
This plastic hopper shelf bin is constructed from durable polypropylene and designed for efficient storage. It features a 50 Lb capacity and measures 17-7/8 in L x 4-3/8 in W x 6 in H, making it suitable for various organizational needs in industrial and laboratory settings. This bin is supplied in a convenient 20-pack and is compatible with a range of shelving units.
  • Molded-in label holder for easy identification.
  • Reinforced edges for enhanced durability.
  • Waterproof and corrosion-free construction ensures longevity.
  • Durable material provides reliable storage.
  • Designed for use with 12 in, 18 in, and 24 in deep shelf units, racks, or standard shelving.
  • Nestable design allows for efficient storage when empty.