Learn More
MSC Quantum Storage Hopper Stacking Bin, Clear, Polypropylene, 8-1/4 x 10-3/4 x 7 in
Catalog No. 5000510597
Encompass
This hopper stacking bin is designed for efficient organization and storage. Made from durable polypropylene, it features a clear design for easy content visibility and offers a 50 lb capacity. Its robust construction allows for stacking and includes features that make it suitable for various demanding environments.
  • Clear styrene window
  • Front, back, and side grip for easy handling
  • Waterproof construction
  • Resistant to extreme cold
  • Resistant to rust and corrosion
  • Autoclavable up to 250 °F
  • Molded ID card slot
  • Stackable for space efficiency
  • Hangable for versatile storage
  • Rectangular shape
Catalog No. 50-005-10597 Supplier MSC Supplier No. 58662784
This hopper stacking bin is designed for efficient organization and storage. Made from durable polypropylene, it features a clear design for easy content visibility and offers a 50 lb capacity. Its robust construction allows for stacking and includes features that make it suitable for various demanding environments.
  • Clear styrene window
  • Front, back, and side grip for easy handling
  • Waterproof construction
  • Resistant to extreme cold
  • Resistant to rust and corrosion
  • Autoclavable up to 250 °F
  • Molded ID card slot
  • Stackable for space efficiency
  • Hangable for versatile storage
  • Rectangular shape