Learn More
MSC Quantum Storage Hopper Stacking Bin, Polypropylene, Blue, 12 Pack
Catalog No. 5000534757
Encompass
Quantum Storage hopper stacking bins are designed for efficient organization and storage. Made from durable polypropylene, these bins are suitable for various industrial and laboratory environments, offering resistance to water, rust, and corrosion.
  • Crystal styrene window
  • FDA approved
  • Water, rust, and corrosion proof
  • Front, back, and side grip
  • Large molded front label
  • Resistant to extreme cold
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F
  • For use with louvered panels or rails
Catalog No. 50-005-34757 Supplier MSC Supplier No. 73087942
Quantum Storage hopper stacking bins are designed for efficient organization and storage. Made from durable polypropylene, these bins are suitable for various industrial and laboratory environments, offering resistance to water, rust, and corrosion.
  • Crystal styrene window
  • FDA approved
  • Water, rust, and corrosion proof
  • Front, back, and side grip
  • Large molded front label
  • Resistant to extreme cold
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F
  • For use with louvered panels or rails