Learn More
MSC Quantum Storage Lid, 23.5 in x 15.5 in, Gray, Polypropylene
Catalog No. 5000534405
Encompass
This gray polypropylene lid is specifically designed for use with Quantum Storage Systems - SNT240 totes, providing a secure closure for your storage containers. Measuring 23.5 inches long, 15.5 inches wide, and 1 inch high, it ensures compatibility and protection for stored items.
  • Designed to fit specific storage totes.
  • Constructed from durable polypropylene.
  • Provides secure closure for storage containers.
  • Measures 23.5 in long, 15.5 in wide, and 1 in high.
  • Order quantity of 1 equals a 3-pack.
Catalog No. 50-005-34405 Supplier MSC Supplier No. 86550746
This gray polypropylene lid is specifically designed for use with Quantum Storage Systems - SNT240 totes, providing a secure closure for your storage containers. Measuring 23.5 inches long, 15.5 inches wide, and 1 inch high, it ensures compatibility and protection for stored items.
  • Designed to fit specific storage totes.
  • Constructed from durable polypropylene.
  • Provides secure closure for storage containers.
  • Measures 23.5 in long, 15.5 in wide, and 1 in high.
  • Order quantity of 1 equals a 3-pack.