Learn More
MSC Quantum Storage Lid, 25.8 x 16.3 x 1.5 in, Blue, Polyethylene
Catalog No. 5000514734
Encompass
This blue lid is designed for use with compatible storage systems. Made from durable polyethylene, it measures 25.8 in long, 16.3 in wide, and 1.5 in high.
  • Constructed from polyethylene
  • Features a blue color
  • Measures 25.8 in long x 16.3 in wide x 1.5 in high
  • Compatible with specific storage tubs
Catalog No. 50-005-14734 Supplier MSC Supplier No. 91826230
This blue lid is designed for use with compatible storage systems. Made from durable polyethylene, it measures 25.8 in long, 16.3 in wide, and 1.5 in high.
  • Constructed from polyethylene
  • Features a blue color
  • Measures 25.8 in long x 16.3 in wide x 1.5 in high
  • Compatible with specific storage tubs