Learn More
MSC Quantum Storage Lid, Gray, Plastic, 25.13 in Length, 16 in Width
Catalog No. 5000536492
Encompass
This gray plastic lid is designed to protect contents in cross-stack tubs, ensuring items remain safe and sanitary during shipping or storage. It effectively shields against dust, dirt, and moisture.
  • Gray color allows for easy product identification.
  • Protects items from dust, dirt, and moisture.
  • Enables cross-stacking for optimal storage.
  • Durable plastic construction for everyday use.
  • Features a snap-on design for ease of use.
  • Made in America.
  • Equipped with a flanged edge.
  • Recessed lip supports 90° cross stacking.
Catalog No. 50-005-36492 Supplier MSC Supplier No. 91826248
This gray plastic lid is designed to protect contents in cross-stack tubs, ensuring items remain safe and sanitary during shipping or storage. It effectively shields against dust, dirt, and moisture.
  • Gray color allows for easy product identification.
  • Protects items from dust, dirt, and moisture.
  • Enables cross-stacking for optimal storage.
  • Durable plastic construction for everyday use.
  • Features a snap-on design for ease of use.
  • Made in America.
  • Equipped with a flanged edge.
  • Recessed lip supports 90° cross stacking.