Learn More
MSC Quantum Storage Mobile Pick Rack Unit, 4,200 lb Capacity, 96 Bins, Ivory
Catalog No. 5000535611
Encompass
This mobile pick rack unit is designed for efficient storage and organization in various industrial and warehouse applications. It features a robust steel construction and provides a high load capacity, making it suitable for heavy-duty use. The unit comes equipped with numerous bins for organized parts storage.
  • Double-sided mobile pick rack unit
  • 4,200 lb load capacity
  • 12 in depth, 75 in height, 36 in width
  • Steel rack material with powder coated finish
  • Includes 96 bins
  • Bins made from polyethylene and polypropylene
  • Bin height is 4 in
  • Ivory bin color
  • 13 shelves with open shelving style
Catalog No. 50-005-35611 Supplier MSC Supplier No. 48520795
This mobile pick rack unit is designed for efficient storage and organization in various industrial and warehouse applications. It features a robust steel construction and provides a high load capacity, making it suitable for heavy-duty use. The unit comes equipped with numerous bins for organized parts storage.
  • Double-sided mobile pick rack unit
  • 4,200 lb load capacity
  • 12 in depth, 75 in height, 36 in width
  • Steel rack material with powder coated finish
  • Includes 96 bins
  • Bins made from polyethylene and polypropylene
  • Bin height is 4 in
  • Ivory bin color
  • 13 shelves with open shelving style